Order Entry
Puerto Rico
ContactUsLinkComponent
Human MMP-8 (from E. coli)
Human MMP-8 (from E. coli)
Catalog # 103010-284
Supplier:  Anaspec
CAS Number:  
Human MMP-8 (from E. coli)
Catalog # 103010-284
Supplier:  Anaspec
Supplier Number:  AS-72008
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µl
  • Storage Conditions:
    –78 °C
  • Protein Synonyms:
    MMP-8 Proenzyme|Matrix metalloproteinase 8|MMP-8 recombinant
  • UniProtKB:
    P22894
  • Protein/Peptide Name:
    MMP-8
  • Molecular Weight:
    85 kDa
  • Concentration:
    10 μg/mL
  • Cat. No.:
    103010-284
  • Supplier no.:
    AS-72008

Specifications

About this item

Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated zinc endopeptidases capable of digesting extracellular matrix components. MMP-8 (neutrophil collagenase) is synthesized by neutrophils and stored in the specific granules until it is secreted.

Human MMP-8 is a full length pro-enzyme isolated from stimulated human neutrophil granulocytes. Proenzyme can be activated by incubating with 1 mM APMA at 37°C for 1 hr. Its activity can be measured in FRET-based enzymatic assays. 10-20 ng of enzyme is sufficient for FRET-based assay.