About this item
Myelin Oligodendrocyte Glycoprotein (MOG) is a member of the immunoglobulin superfamily and is expressed exclusively in the central nervous system (CNS). Although MOG protein constitutes only 0.01-0.05% of the CNS myelin proteins, it was demonstrated that MOG protein is a crucial autoantigen for multiple sclerosis in humans and experimental autoimmune encephalomyelitis (EAE) in rodents and monkeys
The sequence (Accession #CAE84068) corresponding to the extracellular domain of rat MOG along with a 6x His tag was expressed in E. coli. The recombinant rat MOG (R-rMOG) was purified from urea denatured bacterial lysate using immobilized metal affinity chromatography (IMAC). The molecular mass of the recombinant rat MOG is 14.2 kDa.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Rat
- Storage Conditions:–78 °C
- Protein Synonyms:myelin oligodendrocyte protein recombinant|MOG 1-125 recombinant
- Protein/Peptide Name:MOG (1-125)
- Endotoxin Content:Less than 0.1 EU per 1 μg of the protein as determined by Limulus Amebocyte Lysate (LAL) quantitative kinetic assay.
- UniProtKB:Q63345
- Purity:>95% as estimated by SDS-PAGE
- Molecular Weight:14.2 kDa