Specifications
- Antibody Type:Primary
- Antigen Name:breakpoint cluster region
- Antigen Symbol:BCR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg/vial
- Form:Lyophilized
- Gene ID:P11274
- Antigen Synonyms:PHL|D22S11|BCR1|ALL|CML|D22S662
- Molecular Weight:142819
- Storage Temperature:At −20 ˚C for one year. After reconstitution, at 4 ˚C for one month. It can also be aliquotted and stored frozen at −20 ˚C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2 ml of distilled water will yield a concentration of 500 μg/ml
- Shipping Temperature:4 °C
- Immunogen:E.coli-derived human Bcr recombinant protein (Position: M1-A100). Human Bcr shares 94% amino acid (aa) sequence identity with mouse Bcr.
- Purification:Immunogen affinity purified
- Cat. No.:76174-836
Specifications
About this item
Rabbit IgG polyclonal antibody for Breakpoint cluster region protein(BCR) detection. Tested with WB in Human;Mouse;Rat.
1. "Entrez Gene: Breakpoint cluster region".
2. Puil L, Liu J, Gish G, Mbamalu G, Bowtell D, Pelicci PG, Arlinghaus R, Pawson T (February 1994). "Bcr-Abl oncoproteins bind directly to activators of the Ras signalling pathway". EMBO J. 13 (4): 764–73.
3. Pendergast AM, Muller AJ, Havlik MH, Maru Y, Witte ON (July 1991). "BCR sequences essential for transformation by the BCR-ABL oncogene bind to the ABL SH2 regulatory domain in a non-phosphotyrosine-dependent manner".Cell 66 (1): 161–71.
Add 0.2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: BCR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat