Specifications
- Antibody Type:Primary
- Antigen Name:kinesin family member 3A
- Antigen Symbol:KIF3A
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg/vial
- Form:Lyophilized
- Gene ID:Q9Y496
- Antigen Synonyms:FLA10|KLP-20
- UniProtKB:Q9Y496
- Molecular Weight:80041
- Storage Temperature:At −20˚C for one year. After reconstitution, at 4 ˚C for one month. It can also be aliquotted and stored frozen at −20 ˚C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2 ml of distilled water will yield a concentration of 500 μg/ml
- Shipping Temperature:4 °C
- Immunogen:E.coli-derived human KIF3A recombinant protein (Position: D485-Q699). Human KIF3A shares 97.7% amino acid (aa) sequence identity with mouse KIF3A.
- Purification:Immunogen affinity purified
- Cat. No.:76174-164
Specifications
About this item
Rabbit IgG polyclonal antibody for Kinesin-like protein KIF3A(KIF3A) detection. Tested with WB in Human;Mouse;Rat.
Kinesin-like protein KIF3A is a protein that in humans is encoded by the KIF3A gene. KIF3A is one subunit of the heterotrimeric motor protein, kinesin-2, that was initially isolated from sea urchin egg/embryo cytosol using microtubule affinity purification. This motor consists of two kinesin-related subunits (called KIF3A and KIF3B or 3C in vertebrates) and an associated protein (KAP3), and it transports protein complexes, nucleic acids and organelles towards the "plus" ends of microtubule tracks within cells. Work done in a broad range of eukaryotic cells has revealed that heterotrimeric kinesin-2 is the primary motor protein driving the intra-flagellar transport of tubulins and other axonemal building blocks from the base of the ciliary/flagellar axoneme to their site of assembly at the distal tips. This process is required for cilium assembly/maintenance and cilium-based signalling which play key roles in various cell and developmental processes. For example, in vertebrate embryos, kinesin-2 function is required for cilia-dependent nodal flow and the development of left-right asymmetry.
Add 0.2 ml of distilled water will yield a concentration of 500 µg/ml.
Each vial contains 5 mg BSA, 0.9 mg NaCl, 0.2 mg Na₂HPO₄, 0.05 mg NaN₃.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: KIF3A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat