Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ADIPOR1 Rabbit Polyclonal Antibody
Anti-ADIPOR1 Rabbit Polyclonal Antibody
  76173-668
Anti-ADIPOR1 Rabbit Polyclonal Antibody
  76173-668
 :  PB9418

 

  • Antibody Type:
    Primary
  • Antigen Name:
    adiponectin receptor 1
  • Antigen Symbol:
    ADIPOR1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Form:
    Lyophilized
  • Gene ID:
     51094
  • Antigen Synonyms:
    PAQR1|CGI45|CGI-45|TESBP1A|ACDCR1
  • UniProtKB:
    Q96A54
  • Molecular Weight:
    42616
  • Storage Temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Concentration:
    Add 0.2ml of distilled water will yield a concentration of 500 μg/ml.
  • Shipping Temperature:
    4°C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human ADIPOR1 (51-78aa EQTCPVPQEEEEEVRVLTLPLQAHHAME), different from the related mouse sequence by two amino acids.
  • Purification:
    Immunogen affinity purified.
  • Cat. No.:
    76173-668
  • Packaging:
    Vial

 

 

Rabbit IgG polyclonal antibody for Adiponectin receptor protein 1(ADIPOR1) detection. Tested with WB in Human;Rat.

ADIPOR1 is known as Adiponectin receptor protein 1. This gene encodes a protein which acts as a receptor for adiponectin, a hormone secreted by adipocytes which regulates fatty acid catabolism and glucose levels. Binding of adiponectin to the encoded protein results in activation of an AMP-activated kinase signaling pathway which affects levels of fatty acid oxidation and insulin sensitivity. A pseudogene of this gene is located on chromosome 14. Multiple alternatively spliced transcript variants have been found for this gene.

Add 0.2 ml of distilled water will yield a concentration of 500µg/ ml.

Each vial contains 5 mg BSA, 0.9 mg NaCl, 0.2 mg Na2HPO₄, 0.05 mg NaN₃.

WB: The detection limit for ADIPOR1 is approximately 0.1 ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: AHR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat