- Antibody Type:Primary
- Antigen Name:ATP-binding cassette, sub-family G (WHITE), member 5
- Antigen Symbol:ABCG5
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Lyophilized
- Gene ID:64240
- Antigen Synonyms:STSL
- UniProtKB:Q9H222
- Molecular Weight:72504
- Storage Temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2ml of distilled water will yield a concentration of 500 μg/ml.
- Shipping Temperature:4°C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human ABCG5 (197-221aa ERRRVSIAAQLLQDPKVMLFDEPTT), different from the related mouse and rat sequences by two amino acids.
- Purification:Immunogen affinity purified.
- Cat. No.:76173-662
- Packaging:Vial
Rabbit IgG polyclonal antibody for ATP-binding cassette sub-family G member 5(ABCG5) detection. Tested with WB in Human.
ABCG5 (Atp-binding cassette, subfamily g, member 5) also known as STEROLIN 1, is a protein that in humans is encoded by the ABCG5 gene. The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. This protein is a member of the White subfamily. The protein encoded by this gene functions as a half-transporter to limit intestinal absorption and promote biliary excretion of sterols. The ABCG5 gene contains 13 exons and spans about 28 kb. The ABCG5 gene is mapped on 2p21. It is expressed in a tissue-specific manner in the liver, colon, and intestine. This gene is tande mly arrayed on chromosome 2, in a head-to-head orientation with family member ABCG8. Mutations in this gene may contribute to sterol accumulation and atheroschlerosis, and have been observed in patients with sitosterolemia. Small (2003) reviewed the role of ABC transporters in secretion of cholesterol from liver into bile, particularly the role of ABCG5/ABCG8. The ABCG5 and ABCG8 genes are an example of closely neighboring genes in a head-to-head orientation that, when mutated, cause the same phenotype.
Add 0.2 ml of distilled water will yield a concentration of 500ug/ ml.
Each vial contains 5 mg BSA, 0.9 mg NaCl, 0.2 mg Na2HPO₄, 0.05 mg NaN₃.
WB: The detection limit for ABCG5 is approximately 0.1 ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
WB: The detection limit for ABCG5 is approximately 0.1 ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: ACHE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat