Specifications
- Antibody Type:Primary
- Antigen Name:major histocompatibility complex, class I, C
- Antigen Symbol:HLA-C
- Clonality:Polyclonal
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Lyophilized
- Gene ID:3107
- Antigen Synonyms:D6S204|HLC-C|HLA-JY3|PSORS1
- UniProtKB:U5YBW8
- Molecular Weight:8415
- Storage Temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2ml of distilled water will yield a concentration of 500 μg/ml.
- Shipping Temperature:4°C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human HLA-C(38-62aa EYWDRETQKYKRQAQADRVNLRKLR).
- Purification:Immunogen affinity purified.
- Cat. No.:76173-588
- Packaging:Vial
Specifications
About this item
Rabbit IgG polyclonal antibody for MHC class I antigen(HLA-C) detection. Tested with WB, IHC-P, IHC-F, ICC, FCM in Human;Mouse;Rat.
HLA-C belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from endoplasmic reticulum lumen. They are expressed in nearly all cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domain, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. Polymorphisms within exon 2 and exon 3 are responsible for the peptide binding specificity of each class one molecule. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. Over one hundred HLA-C alleles have been described.
Add 0.2 ml of distilled water will yield a concentration of 500µg/ ml.
Each vial contains 5 mg BSA, 0.9 mg NaCl, 0.2 mg Na2HPO₄, 0.05 mg NaN₃.
WB: The detection limit for HLA-C is approximately 0.1 ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: NTRK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
, Human