Specifications
- Antibody Type:Primary
- Antigen Name:caspase 7, apoptosis-related cysteine peptidase
- Antigen Symbol:CASP7
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 µg
- Form:Lyophilized
- Gene ID:840
- Antigen Synonyms:CASP-7|MCH3|ICE-LAP3|CMH-1|LICE2
- UniProtKB:P55210
- Molecular Weight:34277
- Storage Temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2ml of distilled water will yield a concentration of 500 μg/ml.
- Shipping Temperature:4°C
- Immunogen:E.coli-derived human Caspase-7 recombinant protein (Position: A117-D198). Human Caspase-7 shares 92.7% amino acid (aa) sequence identity with both mouse and rat Caspase-7.
- Purification:Immunogen affinity purified.
- Cat. No.:76173-572
- Packaging:Vial
Specifications
About this item
Rabbit IgG polyclonal antibody for Caspase-7(CASP7) detection. Tested with WB, IHC-P in Human;Rat.
CASP7, Caspase-7, apoptosis-related cysteine peptidase, is a human protein encoded by the CASP7 gene. CASP7 orthologs have been identified in nearly all mammals for which complete genome data are available. CASP7 is a member of the caspase (cysteine aspartate protease) family of proteins, and has been shown to be an executioner protein of apoptosis. Using radiation hybrid mapping, the CASP7 gene was localized to human chromosome 10q25.1-q25.2. The orderly activation of CASP7 regulates microglia activation through a protein kinase C-delta (PRKCD)-dependent pathway.
Add 0.2 ml of distilled water will yield a concentration of 500ug/ ml.
Each vial contains 5 mg BSA, 0.9 mg NaCl, 0.2 mg Na2HPO₄, 0.05 mg NaN₃.
WB: The detection limit for Caspase-7 is approximately 0.1 ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: CCND1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human