Specifications
- Antibody Type:Primary
- Antigen Symbol:HSD17B10
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg/vial
- Form:Lyophilized
- Gene ID:Q99714
- Molecular Weight:26923
- Storage Temperature:At −20 ˚C for one year. After reconstitution, at 4 ˚C for one month. It can also be aliquotted and stored frozen at −20 ˚C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2 ml of distilled water will yield a concentration of 500 μg/ml.
- Shipping Temperature:4 °C
- Immunogen:E.coli-derived human ERAB recombinant protein (Position: E48-P261). Human ERAB shares 87% and 88% amino acid (aa) sequence identity with mouse and rat ERAB, respectively.
- Purification:Immunogen affinity purified.
- Cat. No.:76173-506
Specifications
About this item
Rabbit IgG polyclonal antibody for 3-hydroxyacyl-CoA dehydrogenase type-2(HSD17B10) detection. Tested with WB, IHC-P, ICC in Human;Mouse;Rat.
ERAB is also known as HSD17B10 or HADH2. This gene encodes 3-hydroxyacyl-CoA dehydrogenase type II, a member of the short-chain dehydrogenase/reductase superfamily. The gene product is a mitochondrial protein that catalyzes the oxidation of a wide variety of fatty acids and steroids, and is a subunit of mitochondrial ribonuclease P, which is involved in tRNA maturation. The protein has been implicated in the development of Alzheimer disease, and mutations in the gene are the cause of 17beta-hydroxysteroid dehydrogenase type 10 (HSD10) deficiency. Several alternatively spliced transcript variants have been identified, but the full-length nature of only two transcript variants has been determined.
Add 0.2 ml of distilled water will yield a concentration of 500ug/ ml.
Each vial contains 5 mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05 mg NaN3.
WB: The detection limit for ERAB is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: HSPD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat