Specifications
- Antibody Type:Primary
- Antigen Symbol:GRIN2A
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg/vial
- Form:Lyophilized
- Gene ID:Q12879
- Molecular Weight:165283
- Storage Temperature:At −20 ˚C for one year. After reconstitution, at 4 ˚C for one month. It can also be aliquotted and stored frozen at −20 ˚C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2 ml of distilled water will yield a concentration of 500 μg/ml.
- Shipping Temperature:4 °C
- Immunogen:E.coli-derived human NMDAR2A recombinant protein (Position: D958-R1300). Human NMDAR2A shares 89% and 90% amino acid (aa) sequence identity with mouse and rat NMDAR2A, respectively.
- Purification:Immunogen affinity purified.
- Cat. No.:76173-504
Specifications
About this item
Rabbit IgG polyclonal antibody for Glutamate receptor ionotropic, NMDA 2A(GRIN2A) detection. Tested with WB in Human;Mouse;Rat.
GRIN2A is also known as N-methyl-D-aspartate receptor channel, subunit epsilon-1(NMDAR2A). This gene encodes a member of the glutamate-gated ion channel protein family. The encoded protein is an N-methyl-D-aspartate (NMDA) receptor subunit. NMDA receptors are both ligand-gated and voltage-dependent, and are involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. These receptors are permeable to calcium ions, and activation results in a calcium influx into post-synaptic cells, which results in the activation of several signaling cascades. Disruption of this gene is associated with focal epilepsy and speech disorder with or without mental retardation. Alternative splicing results in multiple transcript variants.
Add 0.2 ml of distilled water will yield a concentration of 500 µg/ml.
Each vial contains 5 mg BSA, 0.9mg NaCl, 0.2 mg Na₂HPO₄, 0.05 mg NaN₃.
WB: The detection limit for NMDAR2A is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: HSD17B10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse