Order Entry
United States
ContactUsLinkComponent
Rat Clusterin PicoKine; ELISA Kit, Boster
Rat Clusterin PicoKine; ELISA Kit, Boster
Catalog # 76172-772
CAS Number:  
undefined
Rat Clusterin PicoKine; ELISA Kit, Boster
Catalog # 76172-772
Supplier Number:  EK1615
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Host:
  • Primary antibody reactivity:
  • Target protein:
    Clusterin
  • Description:
    Rat Clusterin PicoKine; ELISA Kit
  • Size:
    96 wells/kit
  • Environmentally Preferable:
  • Cat. No.:
    76172-772

Specifications

About this item

Sandwich High Sensitivity ELISA kit for Quantitative Detection of Rat Clusterin

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Rat Clusterin/Clu with <15pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Clusterin/Clu tissue specificity: Detected in Sertoli cells (at protein level). Detected in cultured Sertoli cells, testis, epididymis, liver and brain.

96wells/kit, with removable strips.