Human IL-23R PicoKine; ELISA Kit, Boster
76172-766
:
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Human
- Target protein:IL-23
- Description:Human IL-23R PicoKine; ELISA Kit
- Size:96 wells/kit
- Cat. No.:76172-766
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Human IL-23R
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Human IL23R with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma(heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. IL23R tissue specificity: Expressed by monocytes, Th1, Th0, NK and dendritic cells. Isoform 1 is specifically expressed in NK cells.
96wells/kit, with removable strips.