Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse TNFRSF19/TROY PicoKine; ELISA Kit, Boster
Mouse TNFRSF19/TROY PicoKine; ELISA Kit, Boster
Catalog # 76172-746
CAS Number:  
undefined
Mouse TNFRSF19/TROY PicoKine; ELISA Kit, Boster
Catalog # 76172-746
Supplier Number:  EK1585
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse TNFRSF19/TROY PicoKine; ELISA Kit
  • Size:
    96 wells/kit
  • Cat. No.:
    76172-746

Specifications

About this item

Sandwich High Sensitivity ELISA kit for Quantitative Detection of Mouse TNFRSF19/TROY

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Mouse Tnfrsf19 with <12pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Tnfrsf19 tissue specificity: Highly expressed in adult brain, and in embryos from day 11-17, but not earlier. Detected in embryonic brain and epithelium, and at lower levels in adult heart, lung and liver. In neonatal mice, mainly in hair follicles and neuron-like cells in the cerebellum, but not in the skin epidermis. Isoform 3 was found in embryonic day 17.5 skin but not in brain and liver.

96wells/kit, with removable strips.