Human Legumain(total) PicoKine; ELISA Kit, Boster
Catalog # 76172-720
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Human
- Description:Human Legumain(total) PicoKine; ELISA Kit
- Size:96 wells/kit
- Cat. No.:76172-720
Specifications
About this item
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Human Legumain(total)
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Human Legumain/LGMN with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Legumain/LGMN tissue specificity: Ubiquitous. Particularly abundant in kidney, heart and placenta.
96wells/kit, with removable strips.