Order Entry
Puerto Rico
ContactUsLinkComponent
Human S100A8/Calgranulin A PicoKine; ELISA Kit, Boster
Human S100A8/Calgranulin A PicoKine; ELISA Kit, Boster
Catalog # 76172-706
CAS Number:  
undefined
Human S100A8/Calgranulin A PicoKine; ELISA Kit, Boster
Catalog # 76172-706
Supplier Number:  EK1558
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Human
  • Description:
    Human S100A8/Calgranulin A PicoKine; ELISA Kit
  • Size:
    96 wells/kit
  • Cat. No.:
    76172-706

Specifications

About this item

Sandwich High Sensitivity ELISA kit for Quantitative Detection of Human S100A8

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Human Calgranulin A/S100A8 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Calgranulin A/S100A8 tissue specificity: Calprotectin (S100A8/9) is predominantly expressed in myeloid cells. Except for inflammatory conditions, the expression is restricted to a specific stage of myeloid differentiation since both proteins are expressed in circulating neutrophils and monocytes but are absent in normal tissue macrophages and lymphocytes. Under chronic inflammatory conditions, such as psoriasis and malignant disorders, also expressed in the epidermis. Found in high concentrations at local sites of inflammation or in the serum of patients with inflammatory diseases such as rheumatoid, cystic fibrosis, inflammatory bowel disease, Crohn's disease, giant cell arteritis, cystic fibrosis, Sjogren's syndrome, systemic lupus erythematosus, and progressive systemic sclerosis. Involved in the formation and deposition of amyloids in the aging prostate known as corpora amylacea inclusions. Strongly up-regulated in many tumors, including gastric, esophageal, colon, pancreatic, bladder, ovarian, thyroid, breast and skin cancers. .

96wells/kit, with removable strips.