- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Human
- Description:Human ADAM15 PicoKine; ELISA Kit
- Size:96 wells/kit
- Cat. No.:76172-584
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Human ADAM15
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Human ADAM15 with <12pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. ADAM15 tissue specificity: Expressed in colon and small intestine. Expressed in airway smooth muscle and glomerular mesangial cells (at protein level). Ubiquitously expressed. Overexpressed in atherosclerotic lesions. Constitutively expressed in cultured endothelium and smooth muscle. Expressed in chondrocytes. Expressed in airway smooth muscle and glomerular mesangial cells. .
96wells/kit, with removable strips.