Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Cripto/TDGF1 PicoKine; ELISA Kit, Boster
Mouse Cripto/TDGF1 PicoKine; ELISA Kit, Boster
Catalog # 76172-500
CAS Number:  
undefined
Mouse Cripto/TDGF1 PicoKine; ELISA Kit, Boster
Catalog # 76172-500
Supplier Number:  EK1405
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse Cripto/TDGF1 PicoKine; ELISA Kit
  • Size:
    96 wells/kit
  • Cat. No.:
    76172-500

Specifications

About this item

Sandwich High Sensitivity ELISA kit for Quantitative Detection of Mouse Cripto/TDGF1

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Mouse Cripto/Tdgf1 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Cripto/Tdgf1 tissue specificity: Expressed at low level in specific organs of the adult animal such as spleen, heart, lung and brain. During gastrulation, expressed in the forming mesoderm. In later stages of the developing heart, expression is restricted to the truncus arteriosus.

96wells/kit, with removable strips.