- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Human
- Description:Human CD97 PicoKine; ELISA Kit
- Size:96 wells/kit
- Cat. No.:76172-496
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Human CD97
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Human CD97 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. CD97 tissue specificity: Broadly expressed, found on most hematopoietic cells, including activated lymphocytes, monocytes, macrophages, dendritic cells, and granulocytes. Expressed also abundantly by smooth muscle cells. Expressed in thyroid, colorectal, gastric, esophageal and pancreatic carcinomas too. Expression are increased under inflammatory conditions in the CNS of multiple sclerosis and in synovial tissue of patients with rheumatoid arthritis. Increased expression of CD97 in the synovium is accompanied by detectable levels of soluble CD97 in the synovial fluid.
96wells/kit, with removable strips.