Order Entry
Puerto Rico
ContactUsLinkComponent
Human Neuregulin-1/NRG1-Beta1 PicoKine; ELISA Kit, Boster
Human Neuregulin-1/NRG1-Beta1 PicoKine; ELISA Kit, Boster
Catalog # 76172-470
CAS Number:  
undefined
Human Neuregulin-1/NRG1-Beta1 PicoKine; ELISA Kit, Boster
Catalog # 76172-470
Supplier Number:  EK1378
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Human
  • Description:
    Human Neuregulin-1/NRG1-Beta1 PicoKine; ELISA Kit
  • Size:
    96 wells/kit
  • Cat. No.:
    76172-470

Specifications

About this item

Sandwich High Sensitivity ELISA kit for Quantitative Detection of Human Neuregulin-1/NRG1-beta1

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Human Neuregulin-1/NRG1 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Neuregulin-1/NRG1 tissue specificity: Type I isoforms are the predominant forms expressed in the endocardium. Isoform alpha is expressed in breast, ovary, testis, prostate, heart, skeletal muscle, lung, placenta liver, kidney, salivary gland, small intestine and brain, but not in uterus, stomach, pancreas, and spleen. Isoform 3 is the predominant form in mesenchymal cells and in non-neuronal organs, whereas isoform 6 is the major neuronal form. Isoform 8 is expressed in spinal cord and brain. Isoform 9 is the major form in skeletal muscle cells; in the nervous system it is expressed in spinal cord and brain. Also detected in adult heart, placenta, lung, liver, kidney, and pancreas. Isoform 10 is expressed in nervous system: spinal cord motor neurons, dorsal root ganglion neurons, and brain. Predominant isoform expressed in sensory and motor neurons. Not detected in adult heart, placenta, lung, liver, skeletal muscle, kidney, and pancreas. Not expressed in fetal lung, liver and kidney. Type IV isoforms are brain-specific. .

96wells/kit, with removable strips.