Order Entry
Puerto Rico
ContactUsLinkComponent
Monkey Primate CXCL14/Brak PicoKine; ELISA Kit, Boster
Monkey Primate CXCL14/Brak PicoKine; ELISA Kit, Boster
Catalog # 76172-422
CAS Number:  
undefined
Monkey Primate CXCL14/Brak PicoKine; ELISA Kit, Boster
Catalog # 76172-422
Supplier Number:  EK1285-PR
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Description:
    Monkey primate CXCL14/Brak PicoKine; ELISA Kit
  • Size:
    96 wells/kit
  • Cat. No.:
    76172-422

Specifications

About this item

Sandwich High Sensitivity ELISA kit for Quantitative Detection of monkey primate CXCL14

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Monkey CXCL14 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma(heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. CXCL14 tissue specificity: Expressed in heart, brain, placenta, lung, liver, skeletal muscle, kidney and pancreas. Highly expressed in normal tissue without inflammatory stimuli and infrequently expressed in cancer cell lines. Weakly expressed in monocyte- derived dendritic cells. Not detected in lung or unstimulated peripheral blood lymphocytes. .

96wells/kit, with removable strips.