Pig porcine CXCL14 PicoKine; ELISA Kit, Boster Biological Technology
Catalog # 76172-420
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Description:Pig porcine CXCL14 PicoKine; ELISA Kit
- Size:96 wells/kit
- Cat. No.:76172-420
Specifications
About this item
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Pig porcine CXCL14
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Pig CXCL14 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma(heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. CXCL14 tissue specificity: Expressed in heart, brain, placenta, lung, liver, skeletal muscle, kidney and pancreas. Highly expressed in normal tissue without inflammatory stimuli and infrequently expressed in cancer cell lines. Weakly expressed in monocyte- derived dendritic cells. Not detected in lung or unstimulated peripheral blood lymphocytes. .
96wells/kit, with removable strips.