- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Human
- Description:Human Tryptase/TPSAB1,B2 PicoKine; ELISA Kit
- Size:96 wells/kit
- Cat. No.:76172-254
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Human Tryptase/TPSAB1,B2
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Human Tryptase Beta-2/TPSB2 with <15pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Tryptase Beta-2/TPSB2 tissue specificity: TPSAB1: Isoform 1 and isoform 2 are expressed in lung,stomach, spleen, heart and skin; in these tissues, isoform 1 ispredominant. Isoform 2 is expressed in aorta, spleen, and breasttumor, with highest levels in the endothelial cells of some bloodvessels surrounding the aorta, as well as those surrounding thetumor and low levels, if any, in mast cells (at protein level).
96wells/kit, with removable strips.