Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse NOV/CCN3 PicoKine; ELISA Kit, Boster
Mouse NOV/CCN3 PicoKine; ELISA Kit, Boster
Catalog # 76172-246
CAS Number:  
undefined
Mouse NOV/CCN3 PicoKine; ELISA Kit, Boster
Catalog # 76172-246
Supplier Number:  EK0834
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse NOV/CCN3 PicoKine; ELISA Kit
  • Size:
    96 wells/kit
  • Cat. No.:
    76172-246

Specifications

About this item

Sandwich High Sensitivity ELISA kit for Quantitative Detection of Mouse NOV/CCN3

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Mouse Ccn3/Nov with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Ccn3/Nov tissue specificity: Expressed in large vessels including the ascending aorta, carotid arteries, and the thoracic aorta, in medium-sized vessels such as coronary arteries and small pulmonary veins and also in small vessels. In addition, also found to be present in the heart (at protein level) (PubMed:21063504). Expressed in astrocytes (at protein level) (PubMed:15213231). Detected in brain, bone, lung and muscle tissues (PubMed:20139355, PubMed:23653360). Expressed in skin, expression highly increases 5 days post-wounding, peaking on the 7th day to decline after 9 days (PubMed:15611078). Expressed in pancreatic ducts and beta-cell islets (PubMed:23705021). .

96wells/kit, with removable strips.