Monkey Primate Survivin PicoKine; ELISA Kit, Boster
Catalog # 76172-176
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Description:Monkey primate Survivin PicoKine; ELISA Kit
- Size:96 wells/kit
- Cat. No.:76172-176
Specifications
About this item
Sandwich High Sensitivity ELISA kit for Quantitative Detection of monkey primate Survivin
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Monkey Survivin/BIRC5 with <2pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma(heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Survivin/BIRC5 tissue specificity: Expressed only in fetal kidney and liver, and to lesser extent, lung and brain. Abundantly expressed in adenocarcinoma (lung, pancreas, colon, breast, and prostate) and in high-grade lymphomas. Also expressed in various renal cell carcinoma cell lines. .
96wells/kit, with removable strips.