Pig porcine TIMP-3 PicoKine; ELISA Kit, Boster Biological Technology
Catalog # 76172-158
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Pig
- Target protein:TIMP-3
- Description:Pig porcine TIMP-3 PicoKine; ELISA Kit
- Size:96 wells/kit
- Cat. No.:76172-158
Specifications
About this item
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Pig porcine TIMP-3
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Pig TIMP3 with <2pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum, plasma(heparin , EDTA) and saliva. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. TIMP3 tissue specificity:
96wells/kit, with removable strips.