Monkey Primate TGF Alpha PicoKine; ELISA Kit, Boster
Catalog # 76172-140
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Monkey
- Target protein:TGF alpha
- Description:Monkey primate TGF Alpha PicoKine; ELISA Kit
- Size:96 wells/kit
- Cat. No.:76172-140
Specifications
About this item
Sandwich High Sensitivity ELISA kit for Quantitative Detection of monkey primate TGF alpha
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Monkey Tgf Alpha/TGFA with <1pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum, plasma(heparin, EDTA) and monkey primate milk. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Tgf Alpha/TGFA tissue specificity: Isoform 1, isoform 3 and isoform 4 are expressed in keratinocytes and tumor-derived cell lines. .
96wells/kit, with removable strips.