Order Entry
Puerto Rico
ContactUsLinkComponent
Monkey Primate NGF/NGF Beta PicoKine; ELISA Kit, Boster
Monkey Primate NGF/NGF Beta PicoKine; ELISA Kit, Boster
Catalog # 76172-114
CAS Number:  
undefined
Monkey Primate NGF/NGF Beta PicoKine; ELISA Kit, Boster
Catalog # 76172-114
Supplier Number:  EK0469-PR
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Description:
    Monkey primate NGF/NGF Beta PicoKine; ELISA Kit
  • Size:
    96 wells/kit
  • Cat. No.:
    76172-114

Specifications

About this item

Sandwich High Sensitivity ELISA kit for Quantitative Detection of monkey primate NGF/NGF beta

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Monkey Beta Ngf/NGF with <1pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates and serum. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Beta Ngf/NGF tissue specificity:

96wells/kit, with removable strips.