Monkey primate BDNF PicoKine; ELISA Kit, Boster
76172-088
:
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Monkey
- Target protein:BDNF
- Description:Monkey primate BDNF PicoKine; ELISA Kit
- Size:96 wells/kit
- Cat. No.:76172-088
Sandwich High Sensitivity ELISA kit for Quantitative Detection of monkey primate BDNF
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Monkey BDNF with <2pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma(heparin, EDTA, citrate). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. BDNF tissue specificity: Brain. Highly expressed in hippocampus, amygdala, cerebral cortex and cerebellum. Also expressed in heart, lung, skeletal muscle, testis, prostate and placenta. .
96wells/kit, with removable strips.