Anti-TNF Alpha Polyclonal Antibody
Catalog # 76173-138
Supplier: Boster Biological Technology
Specifications
- Antibody Type:Primary
- Antigen Name:TNF Alpha
- Antigen Symbol:TNFA
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Rat
- Western Blot:Yes
- Cat. No.:76173-138
Specifications
About this item
Polyclonal antibody for TNF ALPHA/TNF detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Rat. TNF ALPHA/TNF information: Molecular Weight: 25806 MW; Subcellular Localization: Cell membrane ; Single-pass type II membrane protein .
Type: Primary
Antigen: TNF Alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Rat