About this item
Feel the difference of these lightweight gloves, which provide serious protection and secure grip along with the ultimate in dexterity and tactile sensitivity for tasks that demand comfort and control.
- Lightweight nitrile for outstanding dexterity
- Textured fingers for secure handling of wet or oily objects
- Silicone-free for critical environments
- Tear-resistant beaded cuff eases donning
- AQL of 1.0 exceeds laboratory standards
- Consistent quality tip to cuff
TrueForm® is made for those who rely on daily hand protection to provide ease of use and high dexterity. With TrueForm®, quality is designed into every single style using exacting manufacturing processes for tip-to-cuff consistency, glove after glove.
When the need for dexterity and tactile sensitivity are at a premium but the protection of a quality, single-use glove are still a must, the nitrile glove is the preferred choice.
For decades SW® has been the premium manufacturer of single-use performance products. With a relentless focus on new manufacturing technologies and materials that drive innovation, SW® single-use styles continuously push the boundaries of quality and worker-enhancing performance that fuels business success. The SW® difference: 100% air inspected, 75% less residue, consistent tip to cuff.