Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Ammonium acetate 7.5 M, clear solution

Ammonium acetate 7.5 M, clear solution

Ammonium acetate 7.5 M, clear solution
Supplier:  MP Biomedicals
Certificates
Ammonium acetate 7.5 M, clear solution

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Ammonium acetate is a chemical compound which is derived from the reaction of ammonia and acetic acid. Ammonium acetate is volatile at low pressures. Because of which it has been used to replace cell buffers with non-volatile salts, in preparing samples for mass spectrometry. It is used as a buffer for mobile phases for HPLC with ELSD detection.

Ammonium acetate is a widely used reagent in molecular biology and chromatography. Its applications include the purification and precipitation of DNA and protein crystallization. Ammonium acetate is commonly used in HPLC and MS analysis of various compounds, such as oligosaccharides, proteins, and peptides. Ammonium acetate is also used for the nonaqueous capillary electrophoresis-mass spectrometry (NACE-MS) of lipophilic peptides and therapeutic drugs.

Store at +4 °C.

Presentation: Clear Solution

Synonyms:  Acetic acid ammonium salt
  • CAS Number:631-61-8
  • Formula:H₃CCOONH₄
  • MDL Number:MFCD00013066
  • Molecular Weight:77.08 g/mol
  • Density:1.17 (lit.)
  • Storage Temperature:Refrigerator

Documents

Specifications for Product Family

Specification Test Results

  • Identity Test:
    Passes
  • Concentration:
    7.5 M aq (578.25 g/L)
  • Sterility:
    Sterile

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle, amber
    100 ml
    Log in for availability
    Log in for availability
    Each (100ml)
    Each (100ml) - $49.60
    $49.60
    Each (100ml)
       $49.60
    Minimum order quantity is 1

    Supplier Number: 0219875980
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    76180-702
  • Glass bottle, amber
    1 L
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $230.00
    $230.00
    Each (1l)
       $230.00
    Minimum order quantity is 1

    Supplier Number: 0219875991
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    76180-704
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.