Order Entry
Puerto Rico
ContactUsLinkComponent
Glutaraldehyde solution 50% in aqueous solution
Corrosive
Toxic
Irritant
Health
Glutaraldehyde solution 50% in aqueous solution
Supplier:  MP Biomedicals
Certificates
undefined
Corrosive
Toxic
Irritant
Health
Glutaraldehyde solution 50% in aqueous solution
Supplier:  MP Biomedicals

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Identity Test:
    Passes
  • Specific Gravity @ 20/20°C:
    1.126-1.135
  • Glutaraldehyde Assay:
    50%-52% by weight
  • pH @ 25°C:
    3.7-4.5
  • Dilution Haze Test:
    1.0 Transmittance Max
  • Acidity as HOAC:
    <40mL KOH in 0.10N methanolic/60.0g
  • Heavy Metals:
    <0.001%

Specification Test Results