Order Entry
Puerto Rico
ContactUsLinkComponent
Trypan blue 0.4% in PBS, dark blue solution
Trypan blue 0.4% in PBS, dark blue solution
Catalog # 76180-676
Supplier:  MP Biomedicals
CAS Number:  
undefined
Trypan blue 0.4% in PBS, dark blue solution
Catalog # 76180-676
Supplier:  MP Biomedicals
Supplier Number:  091691049
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 ml
  • Cat. No.:
    76180-676

Specifications

Specification Test Results

  • pH:
    7.0-8.0
  • Osmolality:
    270-340mOsm
  • Cell Dye Uptake:
    ≥90%
  • Dye Concentration ((A594± 12nm):
    ≤0.5%

Specification Test Results

About this item

 :
 :960.82 g/mol
CAS Number:72-57-1
Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
MDL Number:MFCD00003969