Order Entry
Puerto Rico
ContactUsLinkComponent
GET Buffer, Rockland Immunochemicals
GET Buffer, Rockland Immunochemicals
Catalog # RLMB-043
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
GET Buffer, Rockland Immunochemicals
Catalog # RLMB-043
Supplier Number:  MB-043
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Specifications

  • Description:
    GET Buffer, 50mM Glucose, 10mM EDTA, 25mM TRIS HCl
  • Size:
    100 mL
  • Cat. No.:
    RLMB-043
  • pH:
    8.0
  • Supplier no.:
    MB-043

Specifications

About this item

GET Buffer pH 8.0, 50 mM Glucose, 10 mM EDTA, 25 mM Tris HCl, pH 8.0 buffer

This product was aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.

Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.