Dionex™ IonPac™ AS11-HC HPLC Column, Thermo Scientific
Catalog # MSPP-052960
Supplier: THERMO ELECTRON NORTH AMERICA MS
Specifications
- Column Format:Analytical
- Flow rate:1.5 – 3.0 mL/min
- Ø int.:4.00 mm
- Particle Size:9 µm
- Pore size:2000 Å
- Description:AS11-HC IC Column
- Length:250 mm
- Column hardware:PEEK
- Cat. No.:MSPP-052960
- Packing material:Ethylvinylbenzene/Divinylbenzene
Specifications
About this item
AS11-HC column used to resolve a larger number of inorganic anions and organic acid anions in complex matrices.
- Higher concentration samples injected without column overloading or peak broadening
- Requires longer retention times, but with higher resolution
- Facilitates rapid analysis in well-characterized samples
- Ideal for complex sample matrices or uncharacterized, for monovalent and divalent organic acids
Designed to resolve analytes in a single run using a hydroxide gradient. The high-capacity column delivers improved separation for monovalent carboxylic acids including quinate, lactate, acetate, propionate, formate, and butyrate.
Complex matrices such as chemical wastewater effluents and fermentation broth solutions contain a variety of inorganic anions and organic acids.