Orders LinkContactUsLinkComponent
Phosphate buffers for microscopy staining, Electron Microscopy Sciences
Phosphate buffers for microscopy staining, Electron Microscopy Sciences
Phosphate buffers for microscopy staining, Electron Microscopy Sciences
Certificates
Phosphate buffers for microscopy staining, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
pH 6.5 is typically used with Wrights stain and potassium ferricyanide solution for Prussian Blue staining of non hemoglobin iron by method of Sundberg and Broman (1955). pH 7.0 buffer is used with Giemsa stain.
Product Family Options
Product Information
  • pH
    Size
    Availability
    Price
  • 101411-138
    Phosphate Buffer
    Phosphate Buffer
    6.5
    500 ml
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $42.95
    $42.95
    Each (500ml)
       $42.95
    Supplier Number: 26161-03
  • 100504-554
    Phosphate Buffer
    Phosphate Buffer
    7.0
    500 ml
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $47.04
    $47.04
    Each (500ml)
       $47.04
    Supplier Number: 26156-02