Pierce™ Streptavidin, Hydrazide-Activated, Thermo Scientific
Catalog # PI21120
Supplier: Invitrogen
CAS Number:
Specifications
- Description:Hydrazide, Activated
- Size:2 mg
- Storage Temperature:Store at room temperature or 4 °C
- Cat. No.:PI21120
- Application:Immunoassay Reagent
- Supplier no.:21120
Specifications
About this item
Pierce™ Hydrazide-activated strepavidin conjugate includes recombinant streptavidin in a purified form activated for crosslinking to carbonyl groups in a molecule.