


Methylcyclohexane 99%, Reagent Grade
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:108-87-2
:
Formula:C₆H₁₁CH₃
MDL Number:MFCD00001497
:98.19 g/mol
Boiling Pt:6.7 %(V)
Melting Pt:–126 °C
Density:0.769 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:203-624-3
UN:2296
ADR:3,II
:GHS07
:GHS08
:GHS02
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 98.5 %
- Vapor Pressure:83.2 mmHg ( 37.7 °C)
- Refractive Index:n20/D 1.422 (lit.)
- Vapor Density:3.4 (vs air)
- Autoignition Temperature:545 °F