Order Entry
Puerto Rico
ContactUsLinkComponent
 
 
 
 
2-Methylbutane ≥99%, Reagent Grade
 
undefined
 
 
 
2-Methylbutane ≥99%, Reagent Grade
 

 

  • Vapor Pressure:
    11.17 psi ( 20 °C)
  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.0 %
  • Autoignition Temperature:
    788 °F
  • Explosion Limit:
    8.3 %
  • Refractive Index:
    n20/D 1.354 (lit.)
  • Vapor Density:
    2.6 (vs air)