


Cyclohexane ≥99.8%, Laboratory Reagent
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:110-82-7
:
Formula:C₆H₁₂
MDL Number:MFCD00003814
:84.16 g/mol
Boiling Pt:8.3 %(V)
Melting Pt:6.4 °C
Density:0.78 g/cm³ (20 °C)
Flash Pt:–18 °C
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
ADR:3,II
:GHS07
:GHS08
:GHS02
Specification Test Results
- Appearance (form):complying
- Appearance (color):complying
- Purity (GC):Min. 99.80 Fl.%
- Non-volatile matter:Max. 0.005 %
- IR spectrum:complying
- Vapor Density:2.9 (vs air)
- Vapor Pressure:77 mmHg ( 20 °C)
- Autoignition Temperature:500 °F
- Refractive Index:n20/D 1.426 (lit.)