Order Entry
United States
ContactUsLinkComponent
N,N-Dimethylformamide ≥99.99995% (trace metals basis), TraceSELECT™ for inorganic trace analysis, Riedel-de Haën™
N,N-Dimethylformamide ≥99.99995% (trace metals basis), TraceSELECT™ for inorganic trace analysis, Riedel-de Haën™
Catalog # BJ72781-1L
CAS Number:  
undefined
N,N-Dimethylformamide ≥99.99995% (trace metals basis), TraceSELECT™ for inorganic trace analysis, Riedel-de Haën™
Catalog # BJ72781-1L
Supplier Number:  72781-1L
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 L
  • Environmentally Preferable:
  • Cat. No.:
    BJ72781-1L

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.8 %
  • Density (D 20/4):
    0.947 - 0.949
  • Silver (Ag):
    Max. 0.5 ppb
  • Aluminium (Al):
    Max. 1 ppb
  • Gold (Au):
    Max. 0.5 ppb
  • Barium (Ba):
    Max. 0.5 ppb
  • Beryllium (Be):
    Max. 0.5 ppb
  • Bismuth (Bi):
    Max. 0.5 ppb
  • Calcium (Ca):
    Max. 20 ppb
  • Cadmium (Cd):
    Max. 0.5 ppb
  • Cerium (Ce):
    Max. 0.5 ppb
  • Cobalt (Co):
    Max. 0.5 ppb
  • Chromium (Cr):
    Max. 1 ppb
  • Cesium (Cs):
    Max. 0.5 ppb
  • Copper (Cu):
    Max. 1 ppb
  • Iron (Fe):
    Max. 5 ppb
  • Gallium (Ga):
    Max. 0.5 ppb
  • Germanium (Ge):
    Max. 0.5 ppb
  • Indium (In):
    Max. 0.5 ppb
  • Iridium (Ir):
    Max. 0.5 ppb
  • Potassium (K):
    Max. 10 ppb
  • Lithium (Li):
    Max. 0.5 ppb
  • Magnesium (Mg):
    Max. 5 ppb
  • Manganese (Mn):
    Max. 0.5 ppb
  • Molybdenum (Mo):
    Max. 0.5 ppb
  • Sodium (Na):
    Max. 200 ppb
  • Nickel (Ni):
    Max. 1 ppb
  • Lead (Pb):
    Max. 0.5 ppb
  • Palladium (Pd):
    Max. 0.5 ppb
  • Platinum (Pt):
    Max. 0.5 ppb
  • Rubidium (Rb):
    Max. 0.5 ppb
  • Rhodium (Rh):
    Max. 0.5 ppb
  • Ruthenium (Ru):
    Max. 0.5 ppb
  • Antimony (Sb):
    Max. 0.5 ppb
  • Tin (Sn):
    Max. 0.5 ppb
  • Strontium (Sr):
    Max. 0.5 ppb
  • Titanium (Ti):
    Max. 1 ppb
  • Thallium (Tl):
    Max. 0.5 ppb
  • Uran (U):
    Max. 0.5 ppb
  • Vanadium (V):
    Max. 0.5 ppb
  • Zinc (Zn):
    Max. 1 ppb
  • Zirconium (Zr):
    Max. 0.5 ppb
  • Explosion Limit:
    15.2 %

Specification Test Results

About this item

Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
CAS Number:68-12-2
 :
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284
 :73.09 g/mol
Boiling Pt:152-153 °C (1.013 hPa)
Boiling Pt:152…153 °C (1.013 hPa)
Melting Pt:-61 °C
Melting Pt:–61 °C
Density:0.94 g/cm³ (20 °C)
Flash Pt:57.5 °C (1.013.25 hPa)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
 :GHS02
 :GHS08
 :GHS07