

Hydrochloric acid 6 M HCl (6N) volumetric solution, Fluka™
Catalog # BJ72033-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 L
- Cat. No.:BJ72033-1L
Specifications
Specification Test Results
- Assay:5.988 - 6.012 mol/l
- Calibrated Against:tris
- Vapor Pressure:190 mmHg ( 25 °C)
Specification Test Results
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Melting Pt:no data available
Density:1.1 g/cm³ (20 °C)
UN:1789
ADR:8,III
:36.46
:GHS07
:GHS05
Storage Temperature:Ambient