
Sodium hydroxide ≥97.0% (by titrimetric analysis), pellets, Purum p.a.
Catalog # BJ71692-5KG
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:5 kg
- Cat. No.:BJ71692-5KG
Specifications
Specification Test Results
- Vapor Pressure:<18 mmHg ( 20 °C)
- Assay:Min. 97 %
- Assay of Na₂CO₃:Max. 0.8 %
- Silver (Ag):Max. 0.001 %
- Aluminium (Al):Max. 0.001 %
- Calcium (Ca):Max. 0.005 %
- Cadmium (Cd):Max. 0.005 %
- Cobalt (Co):Max. 0.005 %
- Copper (Cu):Max. 0.005 %
- Iron (Fe):Max. 0.001 %
- Mercury (Hg):Max. 0.00001 %
- Potassium (K):Max. 0.1 %
- Nickel (Ni):Max. 0.001 %
- Lead (Pb):Max. 0.001 %
- Zinc (Zn):Max. 0.005 %
- Chloride (Cl):Max. 0.005 %
- Total N:Max. 0.0005 %
- Phosphate (PO₄):Max. 0.001 %
- Silicate (as SiO₂):Max. 0.001 %
- Sulfate (SO₄):Max. 0.005 %
Specification Test Results
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:1.390 °C (1.013 hPa)
Melting Pt:319 °C
Density:2.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1823
ADR:8,II
:GHS05