

2-Propanol ≥99.5% ACS, Riedel-de Haën™
Catalog # BJ673773-4L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:4 L
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:EPA Safer chemical solvent (green circle rating); Chem21 solvent selection guide recommended by default and after discussion
- Cat. No.:BJ673773-4L
Specifications
Specification Test Results
- Vapor Pressure:44 mmHg ( 25 °C)
- Identity (IR):complying
- Assay (GC):Min. 99.50 %
- Solubility in water:complying
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.2 %
- Free acid, free alkali:Max. 0.0001 meq/g
- Acetone (GC):Max. 0.002 %
- Carbonyl compounds (as propionaldehyde):Max. 0.002 %
- APHA:Max. 10
- Refractive Index:n20/D 1.377 (lit.)
Specification Test Results
About this item
CAS Number:67-63-0
:
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
:60.1 g/mol
Boiling Pt:77…83 °C (1.013 hPa)
Melting Pt:–90 °C
Density:0.784…0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
:GHS02
:GHS07