Order Entry
Puerto Rico
ContactUsLinkComponent
Ammonium fluoride ≥98.0% eluent additive, for LC-MS, Fluka™
Toxic
Corrosive
Ammonium fluoride ≥98.0% eluent additive, for LC-MS, Fluka™
Catalog # BJ52481-50G
CAS Number:  
Toxic
Corrosive
Ammonium fluoride ≥98.0% eluent additive, for LC-MS, Fluka™
Catalog # BJ52481-50G
Supplier Number:  52481-50G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    50 g
  • Cat. No.:
    BJ52481-50G

Specifications

Specification Test Results

  • Assay:
    98.0 - 102.0 %
  • Ammonium hydrogen difluoride (NH₄HF₂):
    Max. 1 %
  • Residue on ignition (SO₄):
    Max. 0.005 %
  • Silver (Ag):
    Max. 5 ppm
  • Aluminium (Al):
    Max. 5 ppm
  • Barium (Ba):
    Max. 5 ppm
  • Bismuth (Bi):
    Max. 5 ppm
  • Calcium (Ca):
    Max. 50 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 50 ppm
  • Lithium (Li):
    Max. 5 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Molybdenum (Mo):
    Max. 5 ppm
  • Sodium (Na):
    Max. 50 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Strontium (Sr):
    Max. 5 ppm
  • Thallium (Tl):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 50 ppm
  • Suitability for the LC-MS:
    complying

Specification Test Results

About this item

CAS Number:12125-01-8
 :
Formula:NH₄F
MDL Number:MFCD00011423
 :37.04 g/mol
Melting Pt:100 °C
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:235-185-9
UN:2505
ADR:6.1,III
 :GHS06
 :GHS05