
Magnesium 2% in nitric acid 5% ≥99.9995%, TraceSELECT™ matrix modifier solution, Fluka™
Catalog # BJ428892-100ML
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:100 ml
- Cat. No.:BJ428892-100ML
Specifications
Specification Test Results
- Assay of Mg:1.5 - 2.5 %
- Nitric acid:1.0 - 5.0 %
- Metal trace analysis (ICP):Max. 5.5 ppm
Specification Test Results
About this item
:
:24.3 g/mol
Melting Pt:no data available
Density:1.093 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3264
ADR:8,III
:GHS05
CAS Number:7439-95-4
Formula:Mg
MDL Number:MFCD00085308