Order Entry
United States
ContactUsLinkComponent
Methanol ≥99.9%, TraceSELECT™ for metal speciation analysis, Riedel-de Haën™
Methanol ≥99.9%, TraceSELECT™ for metal speciation analysis, Riedel-de Haën™
Catalog # BJ42105-1L
   Safer Human and Environmental Health
CAS Number:  
undefined
Methanol ≥99.9%, TraceSELECT™ for metal speciation analysis, Riedel-de Haën™
Catalog # BJ42105-1L
Supplier Number:  42105-1L
   Safer Human and Environmental Health
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 L
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental Certification:
    Chem21 solvent selection guide recommended after discussion
  • Cat. No.:
    BJ42105-1L

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.9 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Silver (Ag):
    Max. 0.5 ppb
  • Aluminium (Al):
    Max. 5 ppb
  • Arsenic (As):
    Max. 1 ppb
  • Gold (Au):
    Max. 0.5 ppb
  • Barium (Ba):
    Max. 0.5 ppb
  • Beryllium (Be):
    Max. 0.5 ppb
  • Bismuth (Bi):
    Max. 0.5 ppb
  • Calcium (Ca):
    Max. 100 ppb
  • Cadmium (Cd):
    Max. 0.5 ppb
  • Cerium (Ce):
    Max. 0.5 ppb
  • Cobalt (Co):
    Max. 0.5 ppb
  • Chromium (Cr):
    Max. 1 ppb
  • Cesium (Cs):
    Max. 0.5 ppb
  • Copper (Cu):
    Max. 1 ppb
  • Iron (Fe):
    Max. 5 ppb
  • Gallium (Ga):
    Max. 0.5 ppb
  • Germanium (Ge):
    Max. 0.5 ppb
  • Mercury (Hg):
    Max. 1 ppb
  • Indium (In):
    Max. 0.5 ppb
  • Iridium (Ir):
    Max. 0.5 ppb
  • Potassium (K):
    Max. 10 ppb
  • Lithium (Li):
    Max. 0.5 ppb
  • Magnesium (Mg):
    Max. 5 ppb
  • Manganese (Mn):
    Max. 0.5 ppb
  • Molybdenum (Mo):
    Max. 0.5 ppb
  • Sodium (Na):
    Max. 250 ppb
  • Nickel (Ni):
    Max. 2 ppb
  • Lead (Pb):
    Max. 0.5 ppb
  • Palladium (Pd):
    Max. 0.5 ppb
  • Platinum (Pt):
    Max. 0.5 ppb
  • Rubidium (Rb):
    Max. 0.5 ppb
  • Rhodium (Rh):
    Max. 0.5 ppb
  • Ruthenium (Ru):
    Max. 0.5 ppb
  • Antimony (Sb):
    Max. 0.5 ppb
  • Selenium (Se):
    Max. 1 ppb
  • Tin (Sn):
    Max. 0.5 ppb
  • Strontium (Sr):
    Max. 0.5 ppb
  • Titanium (Ti):
    Max. 5 ppb
  • Thallium (Tl):
    Max. 0.5 ppb
  • Uran (U):
    Max. 0.5 ppb
  • Vanadium (V):
    Max. 0.5 ppb
  • Zinc (Zn):
    Max. 50 ppb
  • Zirconium (Zr):
    Max. 0.5 ppb
  • Bromide (Br):
    Max. 10 ppb
  • Chloride (Cl):
    Max. 25 ppb
  • Fluoride (F):
    Max. 10 ppb
  • Iodide (I):
    Max. 10 ppb
  • Nitrite (NO₂):
    Max. 10 ppb
  • Nitrate (NO₃):
    Max. 25 ppb
  • Phosphate (PO₄):
    Max. 10 ppb
  • Sulfate (SO₄):
    Max. 50 ppb
  • Aldehydes (as CH₃CHO):
    Max. 10 ppm
  • Formaldehyde:
    Max. 10 ppm
  • Ketones (as CH₃COCH₃):
    Max. 20 ppm
  • Transmittance at 210 nm:
    Min. 30 %
  • Transmittance at 220 nm:
    Min. 50 %
  • Transmittance at 230 nm:
    Min. 75 %
  • Transmittance from 260 nm:
    Min. 98 %

Specification Test Results

About this item

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
CAS Number:67-56-1
 :
Formula:CH₃OH
MDL Number:MFCD00004595
 :32.04 g/mol
Boiling Pt:64…65 °C (1.013 hPa)
Melting Pt:–98 °C
Density:0.79 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
 :GHS08
 :GHS02