
Silver nitrate concentrate for dilution to 1L, 0.1 M AgNO3 (0.1N) volumetric solution, Fluka™
:7761-88-8
:
:AgNO₃
:MFCD00003414
:169.87 g/mol
:100 °C
:no data available
:Ambient
:3264
:8,II
:GHS05
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective