

Potassium hydroxide concentrate for dilution to 1L, 1.0 M KOH (1.0N) volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Boiling Pt:100 °C
Melting Pt:no data available
Density:1.29…1.37 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1814
ADR:8,II
:GHS07
:GHS05
Specification Test Results
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective