

Acetic acid concentrate for dilution to 1L, 1.0 M CH3COOH (1N) volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
:60.05 g/mol
Boiling Pt:117…118 °C
Melting Pt:4 °C
Flash Pt:19.9 %(V)
Storage Temperature:Ambient
UN:2789
ADR:8,II
:GHS05
:GHS02
Specification Test Results
- Factor:0.998 - 1.002